accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 290-37-9

IUPAC Name: pyrazine
Synonyms: pyrazine paradiazine pyz piazine
Molecular Weight (g/mol): 80.09
Molecular Formula: C4H4N2
InChi Key: KYQCOXFCLRTKLS-UHFFFAOYSA-N
SMILES: C1=CN=CC=N1
Quantity 
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)