accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2902-96-7

IUPAC Name: 2-nitropropan-1-ol
Synonyms: 2-nitropropan-1-ol
Molecular Weight (g/mol): 105.09
Molecular Formula: C3H7NO3
InChi Key: PCNWBUOSTLGPMI-UHFFFAOYNA-N
SMILES: CC(CO)[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results