accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOOClCH3

CAS RN 2905-27-3

IUPAC Name: 3,5-dimethylbenzene-1-sulfonyl chloride
Synonyms: 3,5-dimethylbenzenesulfonyl chloride
Molecular Weight (g/mol): 204.67
Molecular Formula: C8H9ClO2S
InChi Key: LSAGRAXLOLZVKO-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results