accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClN

CAS RN 2905-56-8

IUPAC Name: hydrogen 1-benzylpiperidine chloride
Synonyms: hydrogen 1-benzylpiperidine chloride
Molecular Weight (g/mol): 211.73
Molecular Formula: C12H18ClN
InChi Key: MPPIBJJDFLONMO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C(N1CCCCC1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results