accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOClCl

CAS RN 2905-62-6

IUPAC Name: 3,5-dichlorobenzoyl chloride
Synonyms: 3,5-dichlorobenzoyl chloride
Molecular Weight (g/mol): 209.45
Molecular Formula: C7H3Cl3O
InChi Key: GGHLXLVPNZMBQR-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results