accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOClCl

CAS RN 2905-68-2

IUPAC Name: methyl 3,4-dichlorobenzoate
Synonyms: methyl 3,4-dichlorobenzoate
Molecular Weight (g/mol): 205.03
Molecular Formula: C8H6Cl2O2
InChi Key: WICGATIORFSOJL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Cl)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results