accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29051-44-3

HOON

CAS RN 29051-44-3

IUPAC Name: 6-phenylpyridine-3-carboxylic acid
Synonyms: 6-phenylpyridine-3-carboxylic acid
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: DLFLQXUYRFIFOK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(N=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results