accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29091-40-5

IUPAC Name: 4-methyl-2,1,3-benzoxadiazole
Synonyms: 4-methyl-2,1,3-benzoxadiazole
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: PTXJEYIAUIOTSH-UHFFFAOYSA-N
SMILES: CC1=CC=CC2=NON=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results