accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 291-21-4

IUPAC Name: 1,3,5-trithiane
Synonyms: 1,3,5-trithiane trimethylentrisulfid thioform formaldehyde, thio-, trimer trimethylene trisulfide trithioformaldehyde
Molecular Weight (g/mol): 138.26
Molecular Formula: C3H6S3
InChi Key: LORRLQMLLQLPSJ-UHFFFAOYSA-N
SMILES: C1SCSCS1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results