CAS RN 29110-48-3
IUPAC Name:
hydrogen N-(diaminomethylidene)-2-(2,6-dichlorophenyl)acetamide chloride
Synonyms:
guanfacine
Molecular Weight (g/mol):
282.55
Molecular Formula:
C9H10Cl3N3O
InChi Key:
DGFYECXYGUIODH-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NC(N)=NC(=O)CC1=C(Cl)C=CC=C1Cl
1
–
2
of
2
results
1
Guanfacine Hydrochloride, TRC
CAS: 29110-48-3 Molecular Weight (g/mol): 282.55 Synonym: N-(Aminoiminomethyl)-2,6-dichloro-benzeneacetamide Hydrochloride (1:1),Benzeneacetamide, N-(aminoiminomethyl)-2,6-dichloro-, monohydrochloride (9CI),2,6-Dichlorophenylacetylguanidine hydrochloride,BS 100-141,Estulic,Guanfacine hydrochloride,INTUNIV,LON 798,N-Amidino-2-(2,6-dichlorophenyl)acetamide hydrochloride,SPD 503,Tenex,Tenex (pharmaceutical),[(2,6-Dichlorophenyl)acetyl]guanidine monohydrochloride IUPAC Name: N-carbamimidoyl-2-(2,6-dichlorophenyl)acetamide;hydrochloride SMILES: Cl.NC(=N)NC(=O)Cc1c(Cl)cccc1Cl
2
R&D Systems™ Guanfacine hydrochloride
CAS: 29110-48-3 Molecular Formula: C9H10Cl3N3O