accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29122-68-7

IUPAC Name: 2-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)acetamide
Synonyms: atenolol tensimin myocord atenolol preparation atenol genericon atenol quesada betasyn atenol trom juvental betacard
Molecular Weight (g/mol): 266.34
Molecular Formula: C14H22N2O3
InChi Key: METKIMKYRPQLGS-UHFFFAOYNA-N
SMILES: CC(C)NCC(O)COC1=CC=C(CC(N)=O)C=C1
Molecular Weight (g/mol) 
  • (13)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Industry Type 
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (3)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (13)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)