accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29134-29-0

IUPAC Name: 2-(pyrrolidin-1-yl)acetonitrile
Synonyms: pyrrolidine-1-acetonitrile
Molecular Weight (g/mol): 110.16
Molecular Formula: C6H10N2
InChi Key: NPRYXVXVLCYBNS-UHFFFAOYSA-N
SMILES: N#CCN1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results