accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29198-41-2

IUPAC Name: 1-(1,3-benzothiazol-2-yl)methanamine hydrochloride
Synonyms: 1-(1,3-benzothiazol-2-yl)methanamine hydrochloride
Molecular Weight (g/mol): 200.68
Molecular Formula: C8H9ClN2S
InChi Key: WCZDQDCFSDCIIC-UHFFFAOYSA-N
SMILES: Cl.NCC1=NC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results