accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2921-14-4

IUPAC Name: 2-(aminooxy)acetic acid 2-(azaniumyloxy)acetic acid chloride
Synonyms: (aminooxy)acetic acid (ammoniooxy)acetic acid chloride
Molecular Weight (g/mol): 218.59
Molecular Formula: C4H11ClN2O6
InChi Key: AXIKLJPWEGNZIG-UHFFFAOYSA-N
SMILES: [Cl-].NOCC(O)=O.[NH3+]OCC(O)=O
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)