accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NNNHNHCl

CAS RN 2922-28-3

IUPAC Name: hydrogen 7H-purin-6-amine chloride
Synonyms: hydrogen adenine chloride
Molecular Weight (g/mol): 171.59
Molecular Formula: C5H6ClN5
InChi Key: UQVDQSWZQXDUJB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=C2NC=NC2=NC=N1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results