accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

LiH3COOO

CAS RN 2922-61-4

IUPAC Name: lithium(1+) 2-oxopropanoate
Synonyms: lithium(1+) pyruvate
Molecular Weight (g/mol): 93.99
Molecular Formula: C3H3LiO3
InChi Key: OFJHGWPRBMPXCX-UHFFFAOYSA-M
SMILES: [Li+].CC(=O)C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results