accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29220-04-0

H3CNCH3NSSS

CAS RN 29220-04-0

IUPAC Name: 5-(dimethylamino)-3H-1,2,4-dithiazole-3-thione
Synonyms: 5-(dimethylamino)-1,2,4-dithiazole-3-thione
Molecular Weight (g/mol): 178.29
Molecular Formula: C4H6N2S3
InChi Key: IDRNGWAVAQNFSE-UHFFFAOYSA-N
SMILES: CN(C)C1=NC(=S)SS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results