accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaOOFFF

CAS RN 2923-18-4

IUPAC Name: sodium trifluoroacetate
Synonyms: sodium trifluoroacetate
Molecular Weight (g/mol): 136.01
Molecular Formula: C2F3NaO2
InChi Key: UYCAUPASBSROMS-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C(F)(F)F
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)