accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2923-96-8

IUPAC Name: 4-fluoro-2-nitrobenzaldehyde
Synonyms: 4-fluoro-2-nitrobenzaldehyde
Molecular Weight (g/mol): 169.11
Molecular Formula: C7H4FNO3
InChi Key: ORCGMGUNVGVHDN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results