accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH3CNCH3OPSOH3COH3CN

CAS RN 29232-93-7

IUPAC Name: O-2-(diethylamino)-6-methylpyrimidin-4-yl O,O-dimethyl phosphorothioate
Synonyms: aktel pirimifosmethyl methylpirimiphos epa pesticide chemical code 108102 sybol 2 aktellik pyrimiphos-methyl pirimiphos methyl pirimiphosmethyl aktelik
Molecular Weight (g/mol): 305.33
Molecular Formula: C11H20N3O3PS
InChi Key: QHOQHJPRIBSPCY-UHFFFAOYSA-N
SMILES: CCN(CC)C1=NC(C)=CC(OP(=S)(OC)OC)=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (3)
Solvent or Matrix 
  • (2)
  • (1)
Concentration 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results