accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FF

CAS RN 2927-34-6

IUPAC Name: 1,2-difluoro-4-methylbenzene
Synonyms: 1,2-difluoro-4-methylbenzene
Molecular Weight (g/mol): 128.12
Molecular Formula: C7H6F2
InChi Key: FZMPLKVGINKUJZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(F)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results