accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHOCH3CH3

CAS RN 292870-37-2

IUPAC Name: N-(2,3-dimethylphenyl)-3-methylbenzamide
Synonyms: N-(2,3-dimethylphenyl)-3-methylbenzamide
Molecular Weight (g/mol): 239.32
Molecular Formula: C16H17NO
InChi Key: MTCYJXIELVPXBW-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(=O)NC1=CC=CC(C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results