accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29335-36-2

IUPAC Name: N'-hydroxypropanimidamide
Synonyms: N'-hydroxypropanimidamide
Molecular Weight (g/mol): 88.11
Molecular Formula: C3H8N2O
InChi Key: RLZPCFQNZGINRP-UHFFFAOYSA-N
SMILES: CCC(N)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results