accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONOH3COCH3OH3C

CAS RN 2934-97-6

IUPAC Name: 3,4,10,11-tetramethoxy-7,8,12b,13-tetrahydro-5H-6-azatetraphene
Synonyms: palmatine
Molecular Weight (g/mol): 355.43
Molecular Formula: C21H25NO4
InChi Key: AEQDJSLRWYMAQI-UHFFFAOYNA-N
SMILES: COC1=CC2=C(C=C1OC)C1CC3=C(CN1CC2)C(OC)=C(OC)C=C3
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results