accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2937-50-0

IUPAC Name: prop-2-en-1-yl carbonochloridate
Synonyms: allyl chlorocarbonate
Molecular Weight (g/mol): 120.53
Molecular Formula: C4H5ClO2
InChi Key: CAEWJEXPFKNBQL-UHFFFAOYSA-N
SMILES: ClC(=O)OCC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)