accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 294-90-6

HNNHNHHN

CAS RN 294-90-6

IUPAC Name: 1,4,7,10-tetraazacyclododecane
Synonyms: cyclen
Molecular Weight (g/mol): 172.28
Molecular Formula: C8H20N4
InChi Key: QBPPRVHXOZRESW-UHFFFAOYSA-N
SMILES: C1CNCCNCCNCCN1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)