accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2941-23-3

IUPAC Name: 2-aminocyclopent-1-ene-1-carbonitrile
Synonyms: 2-aminocyclopent-1-ene-1-carbonitrile
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: NSMYBPIHVACKQG-UHFFFAOYSA-N
SMILES: NC1=C(CCC1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)