accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSiOCH3OCH3OCH3

CAS RN 2943-73-9

IUPAC Name: decyltriethoxysilane
Synonyms: decyltriethoxysilane
Molecular Weight (g/mol): 304.55
Molecular Formula: C16H36O3Si
InChi Key: BAAAEEDPKUHLID-UHFFFAOYSA-N
SMILES: CCCCCCCCCC[Si](OCC)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results