accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 295-48-7

CAS RN 295-48-7

IUPAC Name: cyclopentadecane
Synonyms: cyclopentadecane
Molecular Weight (g/mol): 210.41
Molecular Formula: C15H30
InChi Key: SRONXYPFSAKOGH-UHFFFAOYSA-N
SMILES: C1CCCCCCCCCCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results