accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NHOHHCl

CAS RN 29601-98-7

IUPAC Name: N-benzylhydroxylamine hydrochloride
Synonyms: N-benzylhydroxylamine hydrochloride
Molecular Weight (g/mol): 159.61
Molecular Formula: C7H10ClNO
InChi Key: YSNXOQGDHGUKCZ-UHFFFAOYSA-N
SMILES: Cl.ONCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results