accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)ClOO

CAS RN 29617-66-1

IUPAC Name: (2S)-2-chloropropanoate
Synonyms: (2S)-2-chloropropanoate
Molecular Weight (g/mol): 107.51
Molecular Formula: C3H4ClO2
InChi Key: GAWAYYRQGQZKCR-REOHCLBHSA-M
SMILES: C[C@H](Cl)C([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)