accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2IF

CAS RN 29632-74-4

IUPAC Name: 2-fluoro-4-iodoaniline
Synonyms: 2-fluoro-4-iodoaniline
Molecular Weight (g/mol): 237.02
Molecular Formula: C6H5FIN
InChi Key: CUMTUBVTKOYYOU-UHFFFAOYSA-N
SMILES: NC1=CC=C(I)C=C1F
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results