accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3N(S)HO(S)OHNOO

CAS RN 2964-48-9

IUPAC Name: (1S,2S)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-aminium
Synonyms: (1S,2S)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-aminium
Molecular Weight (g/mol): 213.21
Molecular Formula: C9H13N2O4
InChi Key: OCYJXSUPZMNXEN-IUCAKERBSA-O
SMILES: [NH3+][C@@H](CO)[C@@H](O)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)