accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FOOHNOO

CAS RN 29640-98-0

IUPAC Name: 2-(5-fluoro-2-nitrophenyl)acetic acid
Synonyms: (5-fluoro-2-nitrophenyl)acetic acid
Molecular Weight (g/mol): 199.14
Molecular Formula: C8H6FNO4
InChi Key: HOWBVGXZCYNPOU-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(C=CC(F)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results