accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOHHO

CAS RN 29654-55-5

IUPAC Name: 5-(hydroxymethyl)benzene-1,3-diol
Synonyms: benzyl alcohol, 3,5-dihydroxy-
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: NGYYFWGABVVEPL-UHFFFAOYSA-N
SMILES: OCC1=CC(O)=CC(O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results