accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29668-43-7

IUPAC Name: 2,3-dihydro-1,4-benzodioxine-5-carbaldehyde
Synonyms: 2,3-dihydro-1,4-benzodioxine-5-carbaldehyde
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: BJXUCBAQZJITKD-UHFFFAOYSA-N
SMILES: O=CC1=C2OCCOC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results