accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2968-13-0

H3COSF

CAS RN 2968-13-0

IUPAC Name: 1-[(4-fluorophenyl)sulfanyl]propan-2-one
Synonyms: 1-[(4-fluorophenyl)sulfanyl]propan-2-one
Molecular Weight (g/mol): 184.23
Molecular Formula: C9H9FOS
InChi Key: MKJQBSVPZYELIJ-UHFFFAOYSA-N
SMILES: CC(=O)CSC1=CC=C(F)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)