accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29682-15-3

IUPAC Name: methyl 5-bromopyridine-2-carboxylate
Synonyms: methyl 5-bromopyridine-2-carboxylate
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: JEURNBCYNWNADN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Br)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results