accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29682-46-0

IUPAC Name: 1,3-dichloro-2-methyl-4-nitrobenzene
Synonyms: 1,3-dichloro-2-methyl-4-nitrobenzene
Molecular Weight (g/mol): 206.02
Molecular Formula: C7H5Cl2NO2
InChi Key: WBNZUUIFTPNYRN-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results