CAS RN 29741-09-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Apigenin 7-Glucuronide, TRC
CAS: 29741-09-1 Molecular Formula: C21H18O11 Molecular Weight (g/mol): 446.36 Synonym: β-D-5-Hydroxy-2-(p-hydroxyphenyl)-4-oxo-4H-1-benzopyran-7-yl-glucopyranosiduronic Acid,Apigenin 7-O-Glucuronide,Apigenin 7-O-β-D-Glucuronide,Apigenin 7-O-β-D-Glucuronopyranoside,Apigenin 7-O-β-Glucuronide,Apigenin 7-Glucuronide,Apigenin 7-β-Glucuronide,Apigenin-7-O-β-D Glucuronoside,Scutellarin A IUPAC Name: (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[5-hydroxy-2-(4-hydroxyphenyl)-4-oxochromen-7-yl]oxyoxane-2-carboxylic acid SMILES: O[C@H]1[C@H](Oc2cc(O)c3C(=O)C=C(Oc3c2)c4ccc(O)cc4)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O