accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2976-30-9

IUPAC Name: 1-methanesulfonyl-4-nitrobenzene
Synonyms: 1-methanesulfonyl-4-nitrobenzene
Molecular Weight (g/mol): 201.2
Molecular Formula: C7H7NO4S
InChi Key: XONGBDXIFQIQBN-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results