CAS RN 298-03-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Demeton-O, TRC
CAS: 298-03-3 Molecular Formula: C8 H19 O3 P S2 Molecular Weight (g/mol): 258.34 Synonym: S-[2-(Ethylthio)ethyl] O,O-dimethylphosphorothioate,Demetone-O IUPAC Name: diethoxy-(2-ethylsulfanylethoxy)-sulfanylidene-λ^{5}-phosphane SMILES: CCOP(=S)(OCC)OCCSCC