accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 298-07-7

IUPAC Name: bis[(2-ethylhexyl)oxy]phosphinic acid
Synonyms: bis(2-ethylhexyl)phosphate
Molecular Weight (g/mol): 322.43
Molecular Formula: C16H35O4P
InChi Key: SEGLCEQVOFDUPX-UHFFFAOYNA-N
SMILES: CCCCC(CC)COP(O)(=O)OCC(CC)CCCC
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)