accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 298-83-9

IUPAC Name: 2-{3,3'-dimethoxy-4'-[3-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-[1,1'-biphenyl]-4-yl}-3-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Synonyms: nitro blue tetrazolium NBT nitro blue tetrazolium chloride nitroblue tetrazolium chloride nitro blue tetrazolium dichloride
Molecular Weight (g/mol): 817.64
Molecular Formula: C40H30Cl2N10O6
InChi Key: FSVCQIDHPKZJSO-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (9)
  • (2)
Quantity 
  • (1)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Color 
  • (5)
Grade 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (9)
  • (2)
  • (1)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)