accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONONHONOO

CAS RN 2980-33-8

IUPAC Name: 3,5-dinitro-1,2-dihydropyridin-2-one
Synonyms: 3,5-dinitro-1H-pyridin-2-one
Molecular Weight (g/mol): 185.1
Molecular Formula: C5H3N3O5
InChi Key: KSZZJOXNRQKULB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CNC(=O)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results