accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 298709-29-2

IUPAC Name: 3,5-difluoropyridine-2-carbonitrile
Synonyms: 3,5-difluoropyridine-2-carbonitrile
Molecular Weight (g/mol): 140.09
Molecular Formula: C6H2F2N2
InChi Key: WLBIFECTHKFYKV-UHFFFAOYSA-N
SMILES: FC1=CN=C(C#N)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results