accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClMgCl

CAS RN 29874-01-9

IUPAC Name: magnesium(2+) (3-chlorophenyl)methanide chloride
Synonyms: magnesium(2+) (3-chlorophenyl)methanide chloride
Molecular Weight (g/mol): 185.33
Molecular Formula: C7H6Cl2Mg
InChi Key: BQNQIHYLDYHYQP-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].[CH2-]C1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results