accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29886-64-4

IUPAC Name: 4-(thiophen-3-yl)benzoic acid
Synonyms: 4-(thiophen-3-yl)benzoic acid
Molecular Weight (g/mol): 204.24
Molecular Formula: C11H8O2S
InChi Key: FISAUHGRILVMDP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=CSC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)