accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CaHO(R)OH(R)OH(S)OH(R)OHOOHO(R)OH(R)OH(S)OH(R)OHOO

CAS RN 299-28-5

IUPAC Name: calcium bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
Synonyms: calcium gluconate
Molecular Weight (g/mol): 430.37
Molecular Formula: C12H22CaO14
InChi Key: NEEHYRZPVYRGPP-IYEMJOQQSA-L
SMILES: [Ca++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)