CAS RN 299-75-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Treosulfan, TRC
CAS: 299-75-2 Molecular Formula: C6H14O8S2 Molecular Weight (g/mol): 278.3 Synonym: (2S,3S)-1,2,3,4-Butanetetrol 1,4-Dimethanesulfonate,[S-(R*,R*)]-1,2,3,4-Butanetetrol 1,4-Dimethanesulfonate,(2S,3S)-Threitol 1,4-Dimethanesulfonate,L-Threitol 1,4-Bis(methanesulfonate),NSC 39069,Ovastat,Threosulphan,Trecondi,Treosulphan,Tresulfan,(2S,3S)-2,3-Dihydroxybutane-1,4-diyl Dimethanesulfonate IUPAC Name: [(2S,3S)-2,3-dihydroxy-4-methylsulfonyloxybutyl] methanesulfonate SMILES: CS(=O)(=O)OC[C@H](O)[C@@H](O)COS(=O)(=O)C